ADVH49B82AF4 AChemBlock

tert-butyl 2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-8-azaspiro[4.5]dec-2-ene-8-carboxylate

  • Molecular Weight : 363.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-tetramethyl-1-3-2-dioxaborolan-2-yl-8-azaspiro-4-5-dec-2-ene-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84686
Availability details for
Quantity
  • Availability
  • Add to Cart