ADVH49B82AF5 AChemBlock

methyl N-[(3R,4R)-1-benzyl-4-methylpiperidin-3-yl]carbamate

  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-n-3r-4r-1-benzyl-4-methylpiperidin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84878
Availability details for
Quantity
  • Availability
  • Add to Cart