ADVH49B82AF7 AChemBlock

methyl exo-3-hydroxy-7-azabicyclo[2.2.1]heptane-1-carboxylate;hydrochloride

  • Molecular Weight : 207.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-exo-3-hydroxy-7-azabicyclo-2-2-1-heptane-1-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84879
Availability details for
Quantity
  • Availability
  • Add to Cart