ADVH49B82AFE AChemBlock

methyl cis-1-benzyl-4-(hydroxymethyl)pyrrolidine-3-carboxylate

  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-cis-1-benzyl-4-hydroxymethyl-pyrrolidine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84883
Availability details for
Quantity
  • Availability
  • Add to Cart