ADVH49B82B00 AChemBlock

tert-butyl 2-(chloromethyl)-6,7-dihydro-4H-thiazolo[5,4-c]pyridine-5-carboxylate

  • Molecular Weight : 288.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-chloromethyl-6-7-dihydro-4h-thiazolo-5-4-c-pyridine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84688
Availability details for
Quantity
  • Availability
  • Add to Cart