ADVH49B82B05 AChemBlock

Methyl 7-chloro-6-azaindole-3-carboxylate

  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-7-chloro-6-azaindole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72399
Availability details for
Quantity
  • Availability
  • Add to Cart