ADVH49B82B07 AChemBlock

tert-butyl 1-methyl-octahydro-1H-pyrrolo[2,3-c]pyridine-6-carboxylate

  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-methyl-octahydro-1h-pyrrolo-2-3-c-pyridine-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84691
Availability details for
Quantity
  • Availability
  • Add to Cart