ADVH49B82B0B AChemBlock

4,4-difluoro-N-hydroxycyclohexanecarbimidoyl chloride

  • Molecular Weight : 197.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-difluoro-n-hydroxycyclohexanecarbimidoyl-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71904
Availability details for
Quantity
  • Availability
  • Add to Cart