ADVH49B82B17 AChemBlock

methyl 7-(tert-butoxycarbonylamino)-3-azabicyclo[3.2.0]heptane-6-carboxylate

  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-7-tert-butoxycarbonylamino-3-azabicyclo-3-2-0-heptane-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84890
Availability details for
Quantity
  • Availability
  • Add to Cart