ADVH49B82B1F AChemBlock

4-(4-ethylpiperazin-1-yl)-2-nitroaniline

  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-ethylpiperazin-1-yl-2-nitroaniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86459
Availability details for
Quantity
  • Availability
  • Add to Cart