ADVH49B82B20 AChemBlock

4-(4-Chlorophenyl)piperidine hydrochloride

  • Molecular Weight : 232.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-chlorophenyl-piperidine-hydrochloride-293716.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70406
Availability details for
Quantity
  • Availability
  • Add to Cart