ADVH49B82B21 AChemBlock

methyl 6-bromoindolizine-1-carboxylate

  • Molecular Weight : 254.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-bromoindolizine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84894
Availability details for
Quantity
  • Availability
  • Add to Cart