ADVH49B82B22 AChemBlock

4-(3-Trifluoromethyl-phenylsulfanyl)-pyridin-3-ylamine

  • Molecular Weight : 270.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-3-trifluoromethyl-phenylsulfanyl-pyridin-3-ylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86634
Availability details for
Quantity
  • Availability
  • Add to Cart