ADVH49B82B2A AChemBlock

methyl 6-bromo-2-methyl-benzofuran-3-carboxylate

  • Molecular Weight : 269.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-bromo-2-methyl-benzofuran-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84896
Availability details for
Quantity
  • Availability
  • Add to Cart