ADVH49B82B2E AChemBlock

methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazolo[1,5-a]pyridine-3-carboxylate

  • Molecular Weight : 302.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-tetramethyl-1-3-2-dioxaborolan-2-yl-pyrazolo-1-5-a-pyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84899
Availability details for
Quantity
  • Availability
  • Add to Cart