ADVH49B82B38 AChemBlock

3-Iodo-4,6-difluoro indazole

  • Molecular Weight : 280.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-iodo-4-6-difluoro-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72437
Availability details for
Quantity
  • Availability
  • Add to Cart