ADVH49B82B3A AChemBlock

3-fluoro-N-hydroxy-4-(methylcarbamoyl)benzimidoyl chloride

  • Molecular Weight : 230.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-n-hydroxy-4-methylcarbamoyl-benzimidoyl-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71905
Availability details for
Quantity
  • Availability
  • Add to Cart