ADVH49B82B3B AChemBlock

methyl 6-(tert-butoxycarbonylamino)-2-azaspiro[3.3]heptane-7-carboxylate

  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-tert-butoxycarbonylamino-2-azaspiro-3-3-heptane-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84901
Availability details for
Quantity
  • Availability
  • Add to Cart