ADVH49B82B3C AChemBlock

methyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-2-carboxylate

  • Molecular Weight : 301.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-indole-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84902
Availability details for
Quantity
  • Availability
  • Add to Cart