ADVH49B82B41 AChemBlock

tert-butyl 3-ethynylpiperazine-1-carboxylate

  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-ethynylpiperazine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84639
Availability details for
Quantity
  • Availability
  • Add to Cart