ADVH49B82B50 AChemBlock

3-Chloro-4-(morpholine-4-carbonyl)phenylboronic acid pinacol ester

  • Molecular Weight : 369.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-4-morpholine-4-carbonyl-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70863
Availability details for
Quantity
  • Availability
  • Add to Cart