ADVH49B82B52 AChemBlock

methyl 5-bromo-3-methyl-imidazo[1,5-a]pyridine-7-carboxylate

  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-3-methyl-imidazo-1-5-a-pyridine-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84908
Availability details for
Quantity
  • Availability
  • Add to Cart