ADVH49B82B56 AChemBlock

methyl 5,5-difluorooxane-3-carboxylate

  • Molecular Weight : 180.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-5-difluorooxane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84909
Availability details for
Quantity
  • Availability
  • Add to Cart