ADVH49B82B62 AChemBlock

methyl 4-hydroxy-2-oxabicyclo[2.2.2]octane-1-carboxylate

  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-hydroxy-2-oxabicyclo-2-2-2-octane-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84913
Availability details for
Quantity
  • Availability
  • Add to Cart