ADVH49B82B65 AChemBlock

tert-butyl (8R)-8-hydroxy-5-oxa-2-azaspiro[3.5]nonane-2-carboxylate

  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-8r-8-hydroxy-5-oxa-2-azaspiro-3-5-nonane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84699
Availability details for
Quantity
  • Availability
  • Add to Cart