ADVH49B82B6E AChemBlock

methyl 4-bromopyridine-2-carboxylate hydrobromide

  • Molecular Weight : 296.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromopyridine-2-carboxylate-hydrobromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71894
Availability details for
Quantity
  • Availability
  • Add to Cart