ADVH49B82CBC AChemBlock

2-Methylbiphenyl-4-ylboronic acid pinacol ester

  • Molecular Weight : 312.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methylbiphenyl-4-ylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70811
Availability details for
Quantity
  • Availability
  • Add to Cart