ADVH49B82CBF AChemBlock

2-Methoxy-6-isopropyl-3-pyridylboronic acid pinacol ester

  • Molecular Weight : 295.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methoxy-6-isopropyl-3-pyridylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70742
Availability details for
Quantity
  • Availability
  • Add to Cart