ADVH49B82CD4 AChemBlock

2-Bromo-4,5-dimethylphenylboronic acid pinacol ester

  • Molecular Weight : 329.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-4-5-dimethylphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70749
Availability details for
Quantity
  • Availability
  • Add to Cart