ADVH49B82CD6 AChemBlock

2-Benzyloxy-4,5-difluorophenylboronic acid pinacol ester

  • Molecular Weight : 364.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-benzyloxy-4-5-difluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70750
Availability details for
Quantity
  • Availability
  • Add to Cart