ADVH49B82CD7 AChemBlock

2-Benzyloxy-3-chlorophenylboronic acid pinacol ester

  • Molecular Weight : 362.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-benzyloxy-3-chlorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70688
Availability details for
Quantity
  • Availability
  • Add to Cart