ADVH49B82CD8 AChemBlock

2-Benzyloxy-3,4-difluorophenylboronic acid pinacol ester

  • Molecular Weight : 364.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/2-benzyloxy-3-4-difluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70689
Availability details for
Quantity
  • Availability
  • Add to Cart