ADVH49B82CE9 AChemBlock

2,6-Dimethyl-4-isopropoxyphenylboronic acid pinacol ester

  • Molecular Weight : 308.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-6-dimethyl-4-isopropoxyphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70690
Availability details for
Quantity
  • Availability
  • Add to Cart