ADVH49B82D02 AChemBlock

2-(4-Formylphenyl)pyridine-5-boronic acid pinacol ester

  • Molecular Weight : 327.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-4-formylphenyl-pyridine-5-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70534
Availability details for
Quantity
  • Availability
  • Add to Cart