ADVH49B82D73 AChemBlock

(2-Chloro-5-isopropylphenyl)boronic acid pinacol ester

  • Molecular Weight : 298.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-isopropylphenyl-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70497
Availability details for
Quantity
  • Availability
  • Add to Cart