ADVH49B82D75 AChemBlock

(2,2-difluorospiro[2.5]octan-7-yl)methanamine

  • Molecular Weight : 175.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluorospiro-2-5-octan-7-yl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86120
Availability details for
Quantity
  • Availability
  • Add to Cart