ADVH49B82D7F AChemBlock

(1S,2S)-rel-4-amino-2-methyl-cyclohexanecarboxylic acid

  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-2s-rel-4-amino-2-methyl-cyclohexanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86140
Availability details for
Quantity
  • Availability
  • Add to Cart