ADVH49B82D8A AChemBlock

(1S,2R)-rel--4-(tert-butoxycarbonylamino)-2-methyl-cyclohexanecarboxylic acid

  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-2r-rel-4-tert-butoxycarbonylamino-2-methyl-cyclohexanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86154
Availability details for
Quantity
  • Availability
  • Add to Cart