ADVH49B82D8D AChemBlock

(1R,3R,4S)-rel-1-azabicyclo[2.2.1]heptan-3-ol hydrochloride

  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-4s-rel-1-azabicyclo-2-2-1-heptan-3-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86178
Availability details for
Quantity
  • Availability
  • Add to Cart