ADVH49B82D8F AChemBlock

(11S)-3-azaspiro[5.5]undecan-11-amine dihydrochloride

  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/11s-3-azaspiro-5-5-undecan-11-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86195
Availability details for
Quantity
  • Availability
  • Add to Cart