ADVH49B82D92 AChemBlock

(10R)-3-azaspiro[5.5]undecan-10-amine dihydrochloride

  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/10r-3-azaspiro-5-5-undecan-10-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86198
Availability details for
Quantity
  • Availability
  • Add to Cart