ADVH49B82D93 AChemBlock

(1,4-difluorocyclohexyl)methanol

  • Molecular Weight : 150.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-4-difluorocyclohexyl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86199
Availability details for
Quantity
  • Availability
  • Add to Cart