ADVH49B82D94 AChemBlock

(5S,7R,10S,13S,17S)-7-allyl-17-(methoxymethoxy)-10,13-dimethylhexadecahydrospiro[cyclopenta[a]phenanthrene-3,2'-[1,3]dioxolane]

  • Cas Number : 326858-77-9
  • Molecular Weight : 418.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5s-7r-10s-13s-17s-7-allyl-17-methoxymethoxy-10-13-dimethylhexadecahydrospiro-cyclopenta-a-phenanthrene-3-2-1-3-dioxolane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70270
Availability details for
Quantity
  • Availability
  • Add to Cart