ADVH49B82D97 AChemBlock

1-(4-bromoisothiazol-3-yl)-N-methylmethanamine hydrochloride

  • Molecular Weight : 243.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-bromoisothiazol-3-yl-n-methylmethanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71973
Availability details for
Quantity
  • Availability
  • Add to Cart