ADVH49B82D98 AChemBlock

methyl 4-(trifluoromethyl)isothiazole-3-carboxylate

  • Molecular Weight : 211.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-trifluoromethyl-isothiazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72012
Availability details for
Quantity
  • Availability
  • Add to Cart