ADVH49B82D9B AChemBlock

1-(5-fluoroquinolin-8-yl)ethan-1-amine

  • Molecular Weight : 190.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-5-fluoroquinolin-8-yl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72049
Availability details for
Quantity
  • Availability
  • Add to Cart