ADVH49B82D9C AChemBlock

8-(bromomethyl)-3-methylisoquinoline hydrobromide

  • Molecular Weight : 317.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-bromomethyl-3-methylisoquinoline-hydrobromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72055
Availability details for
Quantity
  • Availability
  • Add to Cart