ADVH49B82D9E AChemBlock

4-hydroxy-2-(3-(trifluoromethyl)-3H-diazirin-3-yl)benzaldehyde

  • Molecular Weight : 230.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxy-2-3-trifluoromethyl-3h-diazirin-3-yl-benzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72087
Availability details for
Quantity
  • Availability
  • Add to Cart