ADVH49B82DA0 AChemBlock

5-(trifluoromethyl)imidazo[1,5-a]pyridine

  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-trifluoromethyl-imidazo-1-5-a-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72151
Availability details for
Quantity
  • Availability
  • Add to Cart