ADVH49B82DA4 AChemBlock

(R)-2-chloro-6,7-dihydro-5H-cyclopenta[b]pyridin-7-amine hydrochloride

  • Molecular Weight : 205.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-chloro-6-7-dihydro-5h-cyclopenta-b-pyridin-7-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72172
Availability details for
Quantity
  • Availability
  • Add to Cart